siRNA Details
| Gene Symbol: Cypt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173367.2 | NP_775543.2 | 1 | 229 | ||
| GenBank mRNA | AF463500.1 | - | 1 | 209 | ||
| AK005674.1 | - | 1 | 232 | |||
| AM040452.1 | - | 1 | 224 | |||
| BC048527.1 | - | 1 | 229 | 6772990 | ||

Cypt3
Genomic Map