siRNA Details
| Gene Symbol: Cst12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027054.1 | NP_081330.1 | 2, 3 | 347 | ||
| GenBank mRNA | AF440737.2 | - | 2, 3 | 329 | ||
| AK005670.1 | - | 2, 3 | 338 | |||
| BC119432.2 | - | 2, 3 | 347 | 40041153 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cst12| Gene Symbol: Cst12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027054.1 | NP_081330.1 | 2, 3 | 347 | ||
| GenBank mRNA | AF440737.2 | - | 2, 3 | 329 | ||
| AK005670.1 | - | 2, 3 | 338 | |||
| BC119432.2 | - | 2, 3 | 347 | 40041153 | ||