siRNA Details
| Gene Symbol: 1700012P22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027056.1 | NP_081332.1 | 4 | 600 | ||
| GenBank mRNA | AK005933.1 | - | 4 | 601 | ||
| BC099569.1 | - | 4 | 600 | 30918007 | ||
| BC116372.1 | - | 4 | 526 | 40100233 | ||
| BC126971.1 | - | 4 | 526 | 40100231 | ||

1700012P22Rik
Genomic Map