siRNA Details
| Gene Symbol: Wdfy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111279.1 | NP_001104749.1 | 3 | 320 | ||
| NM_027057.3 | NP_081333.1 | 3 | 320 | |||
| GenBank mRNA | AK005937.1 | - | 3 | 284 | ||
| AK007215.1 | - | 3 | 321 | |||
| AK157204.1 | - | 3 | 279 | |||
| AK159710.1 | - | 3 | 291 | |||
| AK220391.1 | - | 3 | 254 | |||
| BC025226.1 | - | 3 | 234 | 5011404 | ||
| BE197290.1 | - | 3 | 396 | |||

Wdfy1
Genomic Map