siRNA Details
| Gene Symbol: Wdfy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111279.1 | NP_001104749.1 | 5 | 462 | ||
| NM_027057.3 | NP_081333.1 | 5 | 462 | |||
| GenBank mRNA | AK005937.1 | - | 5 | 426 | ||
| AK007215.1 | - | 5 | 463 | |||
| AK157204.1 | - | 5 | 421 | |||
| AK159710.1 | - | 5 | 433 | |||
| AK165554.1 | - | 5 | 434 | |||
| AK190401.1 | - | 2 | 106 | |||
| AK220391.1 | - | 5 | 396 | |||
| BC025226.1 | - | 5 | 376 | 5011404 | ||
| BE197290.1 | - | 1 | 254 | |||

Wdfy1
Genomic Map