siRNA Details
| Gene Symbol: Smco2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027059.1 | NP_081335.1 | 4 | 426 | ||
| GenBank mRNA | AK006257.1 | - | 4 | 433 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smco2| Gene Symbol: Smco2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027059.1 | NP_081335.1 | 4 | 426 | ||
| GenBank mRNA | AK006257.1 | - | 4 | 433 | ||