siRNA Details
| Gene Symbol: 1700024P04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027064.1 | NP_081340.1 | 1 | 219 | ||
| GenBank mRNA | AK006311.1 | - | 1 | 220 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700024P04Rik| Gene Symbol: 1700024P04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027064.1 | NP_081340.1 | 1 | 219 | ||
| GenBank mRNA | AK006311.1 | - | 1 | 220 | ||