siRNA Details
| Gene Symbol: Tmem89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027066.1 | NP_081342.1 | 1 | 222 | ||
| GenBank mRNA | AK006292.1 | - | 1 | 224 | ||
| BC111913.1 | - | 1 | 222 | 40053443 | ||
| BY706371.1 | - | 1 | 224 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem89| Gene Symbol: Tmem89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027066.1 | NP_081342.1 | 1 | 222 | ||
| GenBank mRNA | AK006292.1 | - | 1 | 224 | ||
| BC111913.1 | - | 1 | 222 | 40053443 | ||
| BY706371.1 | - | 1 | 224 | |||