siRNA Details
| Gene Symbol: 1700025G04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_197990.3 | NP_932107.1 | 4 | 724 | ||
| GenBank mRNA | AK006337.1 | - | 5 | 519 | ||
| AK148738.1 | - | 3 | 241 | |||
| AK167392.1 | - | 375 | ||||
| BC034723.1 | - | 4 | 503 | 3981517 | ||
| BC055912.1 | - | 4 | 539 | 3978655 | ||

1700025G04Rik
Genomic Map