siRNA Details
| Gene Symbol: 1700025F22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027074.3 | NP_081350.1 | 4 | 258 | ||
| GenBank mRNA | AK006335.1 | - | 4 | 259 | ||
| BC048529.1 | - | 3 | 186 | 6774736 | ||
| BC061241.1 | - | 4 | 266 | 6743876 | ||

1700025F22Rik
Genomic Map