siRNA Details
| Gene Symbol: Lyzl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027083.1 | NP_081359.1 | 3 | 368 | ||
| GenBank mRNA | AK006279.1 | - | 3 | 285 | ||
| AY566284.1 | - | 2 | 220 | |||
| BC048617.1 | - | 3 | 368 | 6742605 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lyzl6| Gene Symbol: Lyzl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027083.1 | NP_081359.1 | 3 | 368 | ||
| GenBank mRNA | AK006279.1 | - | 3 | 285 | ||
| AY566284.1 | - | 2 | 220 | |||
| BC048617.1 | - | 3 | 368 | 6742605 | ||