siRNA Details
| Gene Symbol: Prss56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027084.1 | NP_081360.1 | 8 | 923 | ||
| GenBank mRNA | JF323950.1 | - | 8 | 923 | ||
| JF698684.1 | - | 8 | 923 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prss56| Gene Symbol: Prss56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027084.1 | NP_081360.1 | 8 | 923 | ||
| GenBank mRNA | JF323950.1 | - | 8 | 923 | ||
| JF698684.1 | - | 8 | 923 | |||