siRNA Details
Gene Symbol: Prss56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027084.1 | NP_081360.1 | 5, 6 | 560 | ||
GenBank mRNA | JF323950.1 | - | 5, 6 | 560 | ||
JF698684.1 | - | 5, 6 | 560 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Prss56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027084.1 | NP_081360.1 | 5, 6 | 560 | ||
GenBank mRNA | JF323950.1 | - | 5, 6 | 560 | ||
JF698684.1 | - | 5, 6 | 560 |