siRNA Details
| Gene Symbol: Clic3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027085.3 | NP_081361.1 | 2 | 128 | ||
| GenBank mRNA | AK009020.1 | - | 2 | 130 | ||
| BC060967.1 | - | 2 | 133 | 30297308 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BC051114.1 | - | 350 | Chr. - - [-] on Build | ||

Clic3
Genomic Map