siRNA Details
| Gene Symbol: Tmem127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175145.3 | NP_780354.1 | 3 | 576 | ||
| GenBank mRNA | AK030324.1 | - | 3 | 597 | ||
| AK050216.1 | - | 3 | 546 | |||
| AK165880.1 | - | 3 | 643 | |||
| BC049832.1 | - | 3 | 576 | 6313802 | ||

Tmem127
Genomic Map