siRNA Details
| Gene Symbol: Etd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006555.1 | - | 2 | 295 | ||
| AY134667.1 | - | 2 | 334 | |||
| BC048661.1 | - | 2 | 248 | 6773570 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Etd| Gene Symbol: Etd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006555.1 | - | 2 | 295 | ||
| AY134667.1 | - | 2 | 334 | |||
| BC048661.1 | - | 2 | 248 | 6773570 | ||