siRNA Details
Gene Symbol: Avpi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027106.4 | NP_081382.1 | 3 | 477 | ||
GenBank mRNA | AK009243.1 | - | 3 | 478 | ||
BC027646.1 | - | 3 | 511 | 1344413 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Avpi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027106.4 | NP_081382.1 | 3 | 477 | ||
GenBank mRNA | AK009243.1 | - | 3 | 478 | ||
BC027646.1 | - | 3 | 511 | 1344413 |