siRNA Details
| Gene Symbol: Ten1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027107.1 | NP_081383.1 | 4 | 575 | ||
| GenBank mRNA | AK009156.1 | - | 4 | 542 | ||
| AK146257.1 | - | 4 | 565 | |||
| BC027639.1 | - | 4 | 520 | 1069092 | ||
| BQ030877.1 | - | 1 | 341 | |||

Ten1
Genomic Map