siRNA Details
| Gene Symbol: 2300002M23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175148.3 | NP_780357.1 | 2 | 647 | ||
| GenBank mRNA | AK037046.1 | - | 2 | 650 | ||
| BC107265.1 | - | 1 | 860 | 40052572 | ||
| BC107266.1 | - | 2 | 647 | 40052573 | ||

2300002M23Rik
Genomic Map