siRNA Details
Gene Symbol: 2310009B15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081226.1 | NP_001074695.1 | 2 | 325 | ||
GenBank mRNA | AK009246.1 | - | 2 | 498 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2310009B15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081226.1 | NP_001074695.1 | 2 | 325 | ||
GenBank mRNA | AK009246.1 | - | 2 | 498 |