siRNA Details
| Gene Symbol: Rhou | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133955.4 | NP_598716.1 | 3 | 499 | ||
| GenBank mRNA | AB051827.1 | - | 3 | 499 | ||
| AF378088.1 | - | 3 | 482 | |||
| BC134407.1 | - | 4 | 336 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rhou| Gene Symbol: Rhou | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133955.4 | NP_598716.1 | 3 | 499 | ||
| GenBank mRNA | AB051827.1 | - | 3 | 499 | ||
| AF378088.1 | - | 3 | 482 | |||
| BC134407.1 | - | 4 | 336 | |||