siRNA Details
| Gene Symbol: Plekhm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033150.1 | NP_001028322.1 | 11 | 2100 | ||
| GenBank mRNA | AK122386.1 | - | 2139 | |||
| AK156652.1 | - | 8 | 1722 | |||
| BC058984.1 | - | 9 | 1691 | |||
| CD355024.1 | - | 2 | 58 | |||

Plekhm2
Genomic Map