siRNA Details
| Gene Symbol: Plekhm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033150.1 | NP_001028322.1 | 4 | 519 | ||
| GenBank mRNA | AK122386.1 | - | 603 | |||
| AK156652.1 | - | 1 | 141 | |||
| AK185623.1 | - | 2 | 109 | |||
| AK219223.1 | - | 3 | 184 | |||
| BC058984.1 | - | 2 | 110 | |||
| CX240883.1 | - | 4 | 519 | |||

Plekhm2
Genomic Map