siRNA Details
| Gene Symbol: 2310061N02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027155.1 | NP_081431.1 | 1 | 552 | ||
| GenBank mRNA | AK010010.1 | - | 1 | 553 | ||
| BC133700.1 | - | 1 | 497 | 40106000 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2310061N02Rik| Gene Symbol: 2310061N02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027155.1 | NP_081431.1 | 1 | 552 | ||
| GenBank mRNA | AK010010.1 | - | 1 | 553 | ||
| BC133700.1 | - | 1 | 497 | 40106000 | ||