siRNA Details
| Gene Symbol: Psmg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101430.2 | NP_001094900.1 | 1 | 150 | ||
| NM_001110515.2 | NP_001103985.1 | 1 | 150 | |||
| GenBank mRNA | AK009887.1 | - | 1 | 35 | ||
| BC070417.1 | - | 1 | 124 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psmg4| Gene Symbol: Psmg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101430.2 | NP_001094900.1 | 1 | 150 | ||
| NM_001110515.2 | NP_001103985.1 | 1 | 150 | |||
| GenBank mRNA | AK009887.1 | - | 1 | 35 | ||
| BC070417.1 | - | 1 | 124 | |||