siRNA Details
| Gene Symbol: Psmg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101430.2 | NP_001094900.1 | 1 | 244 | ||
| NM_001110515.2 | NP_001103985.1 | 1 | 244 | |||
| GenBank mRNA | AA063946.1 | - | 1 | 230 | ||
| AK009887.1 | - | 1 | 129 | |||
| BC070417.1 | - | 1 | 218 | |||

Psmg4
Genomic Map