siRNA Details
Gene Symbol: Ccdc115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027159.2 | NP_081435.1 | 2 | 481 | ||
GenBank mRNA | AK010002.1 | - | 2 | 175 | ||
AK154179.1 | - | 2 | 181 | |||
AK162237.1 | - | 2 | 94 | |||
BC019430.1 | - | 2 | 482 | 3491825 |
