siRNA Details
| Gene Symbol: Tmem52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253856.1 | NP_001240785.1 | 4 | 323 | ||
| NM_001253858.1 | NP_001240787.1 | 3 | 281 | |||
| NM_027161.2 | NP_081437.1 | 4 | 341 | |||
| GenBank mRNA | AK009779.1 | - | 4 | 304 | ||
| BC144992.1 | - | 4 | 340 | 9053540 | ||
| BC150685.1 | - | 4 | 321 | 9087596 | ||

Tmem52
Genomic Map