siRNA Details
| Gene Symbol: Emc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_197991.2 | NP_932108.2 | 5 | 621 | ||
| GenBank mRNA | AK150499.1 | - | 5 | 526 | ||
| AK160274.1 | - | 5 | 538 | |||
| AK169010.1 | - | 5 | 572 | |||
| AK171657.1 | - | 5 | 535 | |||
| AY761096.1 | - | 5 | 514 | |||
| AY761098.1 | - | 5 | 514 | |||
| BC006065.1 | - | 3 | 258 | |||
| BC020179.1 | - | 5 | 516 | 4913500 | ||

Emc10
Genomic Map