siRNA Details
| Gene Symbol: Camsap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163749.1 | NP_001157221.1 | 17 | 3967 | ||
| NM_027171.3 | NP_081447.2 | 17 | 3964 | |||
| GenBank mRNA | AK009960.1 | - | 5 | 551 | ||
| AK081728.1 | - | 17 | 3739 | |||
| AK220401.1 | - | 11 | 4171 | |||
| BC037466.1 | - | 7 | 1802 | |||
| BC043033.1 | - | 9 | 2582 | |||
| BC043081.1 | - | 17 | 3623 | |||
| BC048787.1 | - | 17 | 3612 | 6489361 | ||
| CO042496.1 | - | 1 | 343 | |||
| KC426930.1 | - | 19 | 3686 | |||
| KC426931.1 | - | 19 | 3683 | |||
| KC426932.1 | - | 18 | 3653 | |||
| KC426933.1 | - | 18 | 3650 | |||
| KC426934.1 | - | 18 | 3638 | |||
| KC426935.1 | - | 18 | 3635 | |||
| KC426936.1 | - | 17 | 3605 | |||
| KC426937.1 | - | 17 | 3602 | |||
| KC426938.1 | - | 19 | 2438 | |||
| KC426939.1 | - | 19 | 2435 | |||
| KC426940.1 | - | 18 | 2405 | |||
| KC426941.1 | - | 18 | 2402 | |||
| KC426942.1 | - | 18 | 2390 | |||
| KC426943.1 | - | 18 | 2387 | |||
| KC426944.1 | - | 17 | 2357 | |||
| KC426945.1 | - | 17 | 2354 | |||
| Gene Symbol: Xab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031533.1 | - | 4079 | |||

Camsap3
Genomic Map