siRNA Details
| Gene Symbol: 2310079G19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027173.1 | NP_081449.1 | 1 | 605 | ||
| GenBank mRNA | AK010217.1 | - | 1 | 607 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2310079G19Rik| Gene Symbol: 2310079G19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027173.1 | NP_081449.1 | 1 | 605 | ||
| GenBank mRNA | AK010217.1 | - | 1 | 607 | ||