siRNA Details
| Gene Symbol: Smyd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027188.3 | NP_081464.1 | 10 | 990 | ||
| GenBank mRNA | AK010447.1 | - | 10 | 1024 | ||
| AK043988.1 | - | 10 | 1000 | |||
| AK044168.1 | - | 6 | 551 | |||
| AK046829.1 | - | 7 | 597 | |||
| AK163057.1 | - | 10 | 1007 | |||
| AK167875.1 | - | 10 | 1005 | |||
| BC052431.1 | - | 10 | 990 | 6837012 | ||

Smyd3
Genomic Map