siRNA Details
| Gene Symbol: Tm2d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027194.3 | NP_081470.2 | 4 | 725 | ||
| GenBank mRNA | AK010545.1 | - | 4 | 657 | ||
| AK045768.1 | - | 4 | 684 | |||
| AK080854.1 | - | 4 | 652 | |||
| AK083654.1 | - | 4 | 646 | |||
| AK085975.1 | - | 4 | 677 | |||
| AK086015.1 | - | 4 | 661 | |||
| AK086059.1 | - | 4 | 680 | |||
| AK088246.1 | - | 4 | 1190 | |||
| AK149407.1 | - | 4 | 654 | |||
| AK160039.1 | - | 4 | 662 | |||
| AK166012.1 | - | 4 | 678 | |||
| BC026789.1 | - | 4 | 652 | 4188168 | ||
| CO044843.1 | - | 1 | 570 | |||

Tm2d2
Genomic Map