siRNA Details
| Gene Symbol: 1810046K07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027217.1 | NP_081493.1 | 4 | 272 | ||
| GenBank mRNA | AK007796.1 | - | 4 | 275 | ||
| BC120857.1 | - | 2 | 56 | 40129780 | ||
| BC120859.1 | - | 2 | 56 | 40129782 | ||
 Back To Top
 Back To Top
 1810046K07Rik
1810046K07Rik Genomic Map
Genomic Map Back To Top
 Back To Top