siRNA Details
| Gene Symbol: Prss32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027220.2 | NP_081496.2 | 1 | 50 | ||
| GenBank mRNA | AK008023.1 | - | 1 | 47 | ||
| AY266139.1 | - | 1 | 37 | |||
| BC024903.1 | - | 1 | 50 | 4208033 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prss32| Gene Symbol: Prss32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027220.2 | NP_081496.2 | 1 | 50 | ||
| GenBank mRNA | AK008023.1 | - | 1 | 47 | ||
| AY266139.1 | - | 1 | 37 | |||
| BC024903.1 | - | 1 | 50 | 4208033 | ||