siRNA Details
| Gene Symbol: Vwa9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077631.2 | NP_001071099.1 | 10 | 1494 | ||
| NM_175153.4 | NP_780362.2 | 10 | 1715 | |||
| GenBank mRNA | AK032095.1 | - | 10 | 1709 | ||
| AK035654.1 | - | 10 | 1422 | |||
| AK050654.1 | - | 10 | 1701 | |||
| AK086850.1 | - | 10 | 1408 | |||
| AK168222.1 | - | 9 | 1397 | |||
| AK169772.1 | - | 10 | 1454 | |||

Vwa9
Genomic Map