siRNA Details
| Gene Symbol: Vwa9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077631.2 | NP_001071099.1 | 4 | 743 | ||
| NM_175153.4 | NP_780362.2 | 4 | 964 | |||
| GenBank mRNA | AK032095.1 | - | 4 | 958 | ||
| AK035654.1 | - | 4 | 671 | |||
| AK041578.1 | - | 4 | 659 | |||
| AK050654.1 | - | 4 | 950 | |||
| AK086850.1 | - | 4 | 656 | |||
| AK168222.1 | - | 3 | 646 | |||
| AK169772.1 | - | 4 | 703 | |||
| BC024913.1 | - | 3 | 648 | 3602181 | ||

Vwa9
Genomic Map