siRNA Details
| Gene Symbol: Vwa9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077631.2 | NP_001071099.1 | 5 | 923 | ||
| NM_175153.4 | NP_780362.2 | 5 | 1144 | |||
| GenBank mRNA | AK032095.1 | - | 5 | 1138 | ||
| AK035654.1 | - | 5 | 851 | |||
| AK041578.1 | - | 5 | 839 | |||
| AK050654.1 | - | 5 | 1130 | |||
| AK086850.1 | - | 5 | 837 | |||
| AK168222.1 | - | 4 | 826 | |||
| AK169772.1 | - | 5 | 883 | |||
| BC024913.1 | - | 4 | 828 | 3602181 | ||

Vwa9
Genomic Map