siRNA Details
| Gene Symbol: Zfp219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253694.1 | NP_001240623.1 | 4 | 1305 | ||
| NM_001253695.1 | NP_001240624.1 | 3 | 1034 | |||
| NM_001253696.1 | NP_001240625.1 | 2 | 1103 | |||
| NM_027248.2 | NP_081524.2 | 3 | 1192 | |||
| GenBank mRNA | AB063578.1 | - | 3 | 1192 | ||
| AK008516.1 | - | 1 | 510 | |||
| AK053445.1 | - | 3 | 1031 | |||
| AK154332.1 | - | 4 | 1280 | |||
| BC052050.1 | - | 4 | 1078 | |||
| BC071271.1 | - | 2 | 963 | 30290129 | ||
| BC096486.1 | - | 2 | 1071 | 5352045 | ||

Zfp219
Genomic Map