siRNA Details
| Gene Symbol: Rasip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028544.1 | NP_082820.1 | 7 | 2006 | ||
| GenBank mRNA | AK011557.2 | - | 1 | 89 | ||
| AK033276.1 | - | 5 | 1294 | |||
| AK156602.1 | - | 7 | 2018 | |||
| BC028444.1 | - | 78 | ||||
| BC072584.1 | - | 4 | 960 | |||
| BC150766.1 | - | 7 | 2006 | 9087677 | ||

Rasip1
Genomic Map