siRNA Details
| Gene Symbol: Rab3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023537.5 | NP_076026.1 | 2 | 341 | ||
| GenBank mRNA | AB232588.1 | - | 1 | 192 | ||
| AF312036.1 | - | 1 | 192 | |||
| AK012165.1 | - | 2 | 301 | |||
| AK082959.1 | - | 2 | 341 | |||
| BC051918.1 | - | 2 | 246 | 5691723 | ||
| BC057173.1 | - | 1 | 210 | 4989309 | ||

Rab3b
Genomic Map