siRNA Details
| Gene Symbol: Gm11738 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK039839.1 | - | 2 | 1020 | ||
| Gene Symbol: Dnah17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167746.1 | NP_001161218.1 | 13 | 2040 | ||

Gm11738
Genomic Map