siRNA Details
| Gene Symbol: Dnah17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167746.1 | NP_001161218.1 | 44 | 6646 | ||
| GenBank mRNA | AK173343.1 | - | 2 | 73 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnah17| Gene Symbol: Dnah17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167746.1 | NP_001161218.1 | 44 | 6646 | ||
| GenBank mRNA | AK173343.1 | - | 2 | 73 | ||