siRNA Details
| Gene Symbol: Acot7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146057.1 | NP_001139529.1 | 2 | 274 | ||
| NM_001146058.1 | NP_001139530.1 | 2 | 136 | |||
| NM_133348.2 | NP_579926.2 | 2 | 263 | |||
| GenBank mRNA | AB049821.1 | - | 2 | 229 | ||
| AB088411.1 | - | 2 | 202 | |||
| AB088412.1 | - | 2 | 170 | |||
| AB207243.1 | - | 2 | 229 | |||
| AK152277.1 | - | 2 | 144 | |||
| BC013507.1 | - | 2 | 131 | 4224774 | ||
| BY134190.1 | - | 2 | 276 | |||
| BY327697.1 | - | 2 | 215 | |||
| CA463703.1 | - | 266 | ||||
| CX225643.1 | - | 3 | 167 | |||
| CX235974.1 | - | 2 | 155 | |||

Acot7
Genomic Map