siRNA Details
| Gene Symbol: Dopey2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026700.2 | NP_080976.1 | 3 | 464 | ||
| NM_027293.1 | NP_081569.1 | 3 | 464 | |||
| GenBank mRNA | AK002808.1 | - | 3 | 394 | ||
| AK147255.1 | - | 3 | 442 | |||
| AK147536.1 | - | 2 | 484 | |||
| AK156170.1 | - | 3 | 438 | |||
| AK164839.1 | - | 3 | 441 | |||

Dopey2
Genomic Map