siRNA Details
Gene Symbol: Dancr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012278.1 | - | 3 | 114 | ||
AK045300.1 | - | 858 | ||||
AK172090.1 | - | 2 | 617 | |||
AK172703.1 | - | 1 | 800 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dancr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012278.1 | - | 3 | 114 | ||
AK045300.1 | - | 858 | ||||
AK172090.1 | - | 2 | 617 | |||
AK172703.1 | - | 1 | 800 |