siRNA Details
| Gene Symbol: Ccdc89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027298.1 | NP_081574.1 | 1 | 1222 | ||
| GenBank mRNA | AK006105.1 | - | 1 | 722 | ||
| BC060150.1 | - | 1 | 956 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc89| Gene Symbol: Ccdc89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027298.1 | NP_081574.1 | 1 | 1222 | ||
| GenBank mRNA | AK006105.1 | - | 1 | 722 | ||
| BC060150.1 | - | 1 | 956 | |||