siRNA Details
| Gene Symbol: Nol7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023554.2 | NP_076043.2 | 2 | 329 | ||
| GenBank mRNA | AK008670.1 | - | 2 | 314 | ||
| AK019972.1 | - | 2 | 314 | |||
| AK032963.1 | - | 2 | 315 | |||
| AK207964.1 | - | 2 | 84 | |||
| BC090637.1 | - | 2 | 302 | 30940523 | ||
| BC130012.1 | - | 2 | 319 | 40140312 | ||
| BY099842.1 | - | 2 | 329 | |||

Nol7
Genomic Map