siRNA Details
| Gene Symbol: Igsf23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027308.2 | NP_081584.1 | 3 | 553 | ||
| GenBank mRNA | AK008696.1 | - | 3 | 555 | ||
| AK033677.1 | - | 3 | 554 | |||
| AK088927.1 | - | 4 | 509 | |||
| AK135657.1 | - | 359 | ||||
| BC139455.1 | - | 3 | 401 | 8862477 | ||
| BC139458.1 | - | 3 | 401 | 8862480 | ||

Igsf23
Genomic Map