siRNA Details
| Gene Symbol: Odf3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013022.1 | NP_001013040.1 | 5 | 634 | ||
| GenBank mRNA | AK008010.1 | - | 4 | 526 | ||
| BC087962.1 | - | 5 | 634 | 6748887 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Odf3b| Gene Symbol: Odf3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013022.1 | NP_001013040.1 | 5 | 634 | ||
| GenBank mRNA | AK008010.1 | - | 4 | 526 | ||
| BC087962.1 | - | 5 | 634 | 6748887 | ||